2-(Isobutyrylamino)acetic acid

Supplier: Matrix Scientific

049268-500MG 049268-2.5G
101886-976EA 210.77 USD
101886-976 101886-978
2-(Isobutyrylamino)acetic acid
2-(Isobutyrylamino)acetic acid

Hazard: Irritant

Formula: C₆H₁₁NO₃
MDL Number: MFCD09043666
CAS Number: 15926-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR