3-Pyrrolidinyl-2-methylpropanoate hydrochloride
Supplier: Matrix Scientific
049265-500MG
049265-2.5G
101886-964EA
364.62
USD
101886-964
101886-966
3-Pyrrolidinyl-2-methylpropanoate hydrochloride
3-Pyrrolidinyl-2-methylpropanoate hydrochloride
Hazard: Irritant
Formula:
C₈H₁₆ClNO₂ MW: 193.68 g/mol |
MDL Number:
MFCD13559786 CAS Number: 1219841-63-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...