2-(2-Piperidinyl)ethyl-2-methylpropanoate hydrochloride

Supplier: Matrix Scientific

049263-500MG 049263-2.5G
101886-956EA 364.62 USD
101886-956 101886-958
2-(2-Piperidinyl)ethyl-2-methylpropanoate hydrochloride
2-(2-Piperidinyl)ethyl-2-methylpropanoate hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₂ClNO₂
MW: 235.76 g/mol
MDL Number: MFCD13559784
CAS Number: 1219979-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR