2-(3-Piperidinyl)ethyl-2-methylpropanoate hydrochloride
Supplier: Matrix Scientific
049262-500MG
049262-2.5G
101886-952EA
364.62
USD
101886-952
101886-954
2-(3-Piperidinyl)ethyl-2-methylpropanoate hydrochloride
2-(3-Piperidinyl)ethyl-2-methylpropanoate hydrochloride
Hazard: Irritant
Formula:
C₁₁H₂₂ClNO₂ |
MDL Number:
MFCD13559783 CAS Number: 1220032-00-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...