3-Piperidinylmethyl-2-methylpropanoate hydrochloride

Supplier: Matrix Scientific

049259-500MG 049259-2.5G
101886-940EA 364.62 USD
101886-940 101886-942
3-Piperidinylmethyl-2-methylpropanoate hydrochloride
3-Piperidinylmethyl-2-methylpropanoate hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀ClNO₂
MDL Number: MFCD13559780
CAS Number: 1219979-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR