3-Azetidinyl-2-methylpropanoate
Supplier: Matrix Scientific
049258-500MG
049258-2.5G
101886-936EA
364.62
USD
101886-936
101886-938
3-Azetidinyl-2-methylpropanoate
3-Azetidinyl-2-methylpropanoate
Hazard: Irritant
Formula:
C₇H₁₃NO₂ |
MDL Number:
MFCD13559779 CAS Number: 1220031-99-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...