4-Piperidinyl-2-methylpropanoate hydrochloride

Supplier: Matrix Scientific

049257-500MG 049257-2.5G
101886-932EA 364.62 USD
101886-932 101886-934
4-Piperidinyl-2-methylpropanoate hydrochloride
4-Piperidinyl-2-methylpropanoate hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO₂
MW: 207.7 g/mol
MDL Number: MFCD13559778
CAS Number: 1219980-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR