3-Piperidinyl-2-methylpropanoate hydrochloride

Supplier: Matrix Scientific

049256-500MG 049256-2.5G
101886-928EA 364.62 USD
101886-928 101886-930
3-Piperidinyl-2-methylpropanoate hydrochloride
3-Piperidinyl-2-methylpropanoate hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO₂
MW: 207.7 g/mol
MDL Number: MFCD13559777
CAS Number: 1220020-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR