1-Propionyl-2-piperidinecarboxylic acid

Supplier: Matrix Scientific

049251-500MG 049251-2.5G
101886-908EA 364.62 USD
101886-908 101886-910
1-Propionyl-2-piperidinecarboxylic acid
1-Propionyl-2-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₉H₁₅NO₃
MDL Number: MFCD09046790
CAS Number: 1103289-56-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR