N-(4-Piperidinyl)propanamide hydrochloride
Supplier: Matrix Scientific
049249-500MG
049249-2.5G
101886-900EA
364.62
USD
101886-900
101886-902
N-(4-Piperidinyl)propanamide hydrochloride
N-(4-Piperidinyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₈H₁₇ClN₂O |
MDL Number:
MFCD11809619 CAS Number: 183732-59-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...