3-Pyrrolidinylmethyl propanoate hydrochloride

Supplier: Matrix Scientific

049246-500MG 049246-2.5G
101886-888EA 364.62 USD
101886-888 101886-890
3-Pyrrolidinylmethyl propanoate hydrochloride
3-Pyrrolidinylmethyl propanoate hydrochloride

Hazard: Irritant

Formula: C₈H₁₆ClNO₂
MDL Number: MFCD13559773
CAS Number: 1220021-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR