2-(2-Piperidinyl)ethyl propanoate hydrochloride
Supplier: Matrix Scientific
049245-500MG
049245-2.5G
101886-884EA
364.62
USD
101886-884
101886-886
2-(2-Piperidinyl)ethyl propanoate hydrochloride
2-(2-Piperidinyl)ethyl propanoate hydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₀ClNO₂ |
MDL Number:
MFCD13559772 CAS Number: 1220021-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...