2-(4-Piperidinyl)ethyl propanoate hydrochloride
Supplier: Matrix Scientific
049243-500MG
049243-2.5G
101886-876EA
364.62
USD
101886-876
101886-878
2-(4-Piperidinyl)ethyl propanoate hydrochloride
2-(4-Piperidinyl)ethyl propanoate hydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₀ClNO₂ |
MDL Number:
MFCD13559770 CAS Number: 184044-16-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...