4-Piperidinylmethyl propanoate hydrochloride

Supplier: Matrix Scientific

049242-500MG 049242-2.5G
101886-872EA 364.62 USD
101886-872 101886-874
4-Piperidinylmethyl propanoate hydrochloride
4-Piperidinylmethyl propanoate hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO₂
MDL Number: MFCD13559769
CAS Number: 184042-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR