3-Piperidinyl propanoate hydrochloride

Supplier: Matrix Scientific

049238-500MG 049238-2.5G
101886-856EA 364.62 USD
101886-856 101886-858
3-Piperidinyl propanoate hydrochloride
3-Piperidinyl propanoate hydrochloride

Hazard: Irritant

Formula: C₈H₁₆ClNO₂
MDL Number: MFCD13559766
CAS Number: 1220031-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR