N-(4-Methoxybenzoyl)-2-methylalanine

Supplier: Matrix Scientific

049235-500MG 049235-2.5G
101886-844EA 364.62 USD
101886-844 101886-846
N-(4-Methoxybenzoyl)-2-methylalanine
N-(4-Methoxybenzoyl)-2-methylalanine

Hazard: Irritant

Formula: C₁₂H₁₅NO₄
MDL Number: MFCD13325375
CAS Number: 38388-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR