1-(4-Methoxybenzoyl)-3-piperidinecarboxylic acid

Supplier: Matrix Scientific

049234-500MG 049234-2.5G
101886-840EA 364.62 USD
101886-840 101886-842
1-(4-Methoxybenzoyl)-3-piperidinecarboxylic acid
1-(4-Methoxybenzoyl)-3-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₇NO₄
MDL Number: MFCD04035218
CAS Number: 436093-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR