1-(4-Methoxybenzoyl)-2-piperidinecarboxylic acid

Supplier: Matrix Scientific

049233-500MG 049233-2.5G
101886-836EA 364.62 USD
101886-836 101886-838
1-(4-Methoxybenzoyl)-2-piperidinecarboxylic acid
1-(4-Methoxybenzoyl)-2-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₇NO₄
MDL Number: MFCD09050426
CAS Number: 1103529-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR