N-(4-Methoxybenzoyl)alanine

Supplier: Matrix Scientific

049231-500MG 049231-2.5G
101886-828EA 364.62 USD
101886-828 101886-830
N-(4-Methoxybenzoyl)alanine
N-(4-Methoxybenzoyl)alanine

Hazard: Irritant

Formula: C₁₁H₁₃NO₄
MDL Number: MFCD09757664
CAS Number: 93709-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR