4-Methoxy-N-(4-piperidinyl)benzamide hydrochloride
Supplier: Matrix Scientific
049230-500MG
049230-2.5G
101886-824EA
364.62
USD
101886-824
101886-826
4-Methoxy-N-(4-piperidinyl)benzamide hydrochloride
4-Methoxy-N-(4-piperidinyl)benzamide hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉ClN₂O₂ |
MDL Number:
MFCD13559765 CAS Number: 1219948-56-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...