Methyl (2S,4S)-4-[(4-methoxybenzoyl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

049229-500MG 049229-2.5G
101886-820EA 364.62 USD
101886-820 101886-822
Methyl (2S,4S)-4-[(4-methoxybenzoyl)oxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[(4-methoxybenzoyl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₈ClNO₅
MDL Number: MFCD13559764
CAS Number: 1354488-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR