3-Pyrrolidinyl-4-methoxybenzoate hydrochloride

Supplier: Matrix Scientific

049228-500MG 049228-2.5G
101886-816EA 364.62 USD
101886-816 101886-818
3-Pyrrolidinyl-4-methoxybenzoate hydrochloride
3-Pyrrolidinyl-4-methoxybenzoate hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆ClNO₃
MDL Number: MFCD13559763
CAS Number: 1220021-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR