3-Pyrrolidinylmethyl-4-methoxybenzoate hydrochloride

Supplier: Matrix Scientific

049227-500MG 049227-2.5G
101886-812EA 364.62 USD
101886-812 101886-814
3-Pyrrolidinylmethyl-4-methoxybenzoate hydrochloride
3-Pyrrolidinylmethyl-4-methoxybenzoate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClNO₃
MDL Number: MFCD13559762
CAS Number: 1220037-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR