2-(2-Piperidinyl)ethyl-4-methoxybenzoate hydrochloride
Supplier: Matrix Scientific
049226-500MG
049226-2.5G
101886-808EA
364.62
USD
101886-808
101886-810
2-(2-Piperidinyl)ethyl-4-methoxybenzoate hydrochloride
2-(2-Piperidinyl)ethyl-4-methoxybenzoate hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₂ClNO₃ |
MDL Number:
MFCD13559761 CAS Number: 1220031-97-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...