2-(4-Piperidinyl)ethyl-4-methoxybenzoate hydrochloride
Supplier: Matrix Scientific
049224-500MG
049224-2.5G
101886-800EA
364.62
USD
101886-800
101886-802
2-(4-Piperidinyl)ethyl-4-methoxybenzoate hydrochloride
2-(4-Piperidinyl)ethyl-4-methoxybenzoate hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₂ClNO₃ MW: 299.8 g/mol |
MDL Number:
MFCD13559759 CAS Number: 1220021-18-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...