4-Piperidinylmethyl-4-methoxybenzoate hydrochloride

Supplier: Matrix Scientific

049223-500MG 049223-2.5G
101886-796EA 364.62 USD
101886-796 101886-798
4-Piperidinylmethyl-4-methoxybenzoate hydrochloride
4-Piperidinylmethyl-4-methoxybenzoate hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀ClNO₃
MDL Number: MFCD13559758
CAS Number: 1220037-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR