3-Piperidinylmethyl-4-methoxybenzoate hydrochloride

Supplier: Matrix Scientific

049222-500MG 049222-2.5G
101886-792EA 364.62 USD
101886-792 101886-794
3-Piperidinylmethyl-4-methoxybenzoate hydrochloride
3-Piperidinylmethyl-4-methoxybenzoate hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀ClNO₃
MDL Number: MFCD13559757
CAS Number: 1220031-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR