3-Azetidinyl-4-methoxybenzoate

Supplier: Matrix Scientific

049221-500MG 049221-2.5G
101886-788EA 364.62 USD
101886-788 101886-790
3-Azetidinyl-4-methoxybenzoate
3-Azetidinyl-4-methoxybenzoate

Hazard: Irritant

Formula: C₁₁H₁₃NO₃
MDL Number: MFCD13559756
CAS Number: 1220021-08-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR