3-Piperidinyl-4-methoxybenzoate hydrochloride

Supplier: Matrix Scientific

049219-500MG 049219-2.5G
101886-780EA 364.62 USD
101886-780 101886-782
3-Piperidinyl-4-methoxybenzoate hydrochloride
3-Piperidinyl-4-methoxybenzoate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClNO₃
MDL Number: MFCD13559754
CAS Number: 1220031-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR