N-(Tetrahydro-2H-pyran-4-ylcarbonyl)-β-alanine

Supplier: Matrix Scientific

049218-500MG 049218-2.5G
101886-776EA 364.62 USD
101886-776 101886-778
N-(Tetrahydro-2H-pyran-4-ylcarbonyl)-β-alanine
N-(Tetrahydro-2H-pyran-4-ylcarbonyl)-beta-alanine

Hazard: Irritant

Formula: C₉H₁₅NO₄
MW: 201.22 g/mol
MDL Number: MFCD11170003
CAS Number: 1042797-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR