2-[Methyl(tetrahydro-2H-pyran-4-ylcarbonyl)amino]acetic acid

Supplier: Matrix Scientific

049217-500MG 049217-2.5G
101886-772EA 364.62 USD
101886-772 101886-774
2-[Methyl(tetrahydro-2H-pyran-4-ylcarbonyl)amino]acetic acid
2-[Methyl(tetrahydro-2H-pyran-4-ylcarbonyl)amino]acetic acid

Hazard: Irritant

Formula: C₉H₁₅NO₄
MDL Number: MFCD11170103
CAS Number: 1042800-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR