2-Methyl-N-(tetrahydro-2H-pyran-4-ylcarbonyl)alanine

Supplier: Matrix Scientific

049216-500MG 049216-2.5G
101886-768EA 364.62 USD
101886-768 101886-770
2-Methyl-N-(tetrahydro-2H-pyran-4-ylcarbonyl)alanine
2-Methyl-N-(tetrahydro-2H-pyran-4-ylcarbonyl)alanine

Hazard: Irritant

Formula: C₁₀H₁₇NO₄
MW: 215.25 g/mol
MDL Number: MFCD13559753
CAS Number: 1219979-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR