1-(Tetrahydro-2H-pyran-4-ylcarbonyl)-4-piperidinecarboxylic acid

Supplier: Matrix Scientific

049215-500MG 049215-2.5G
101886-764EA 364.62 USD
101886-764 101886-766
1-(Tetrahydro-2H-pyran-4-ylcarbonyl)-4-piperidinecarboxylic acid
1-(Tetrahydro-2H-pyran-4-ylcarbonyl)-4-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₉NO₄
MDL Number: MFCD11169963
CAS Number: 1042796-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR