1-(Tetrahydro-2H-pyran-4-ylcarbonyl)-3-piperidinecarboxylic acid

Supplier: Matrix Scientific

049214-500MG 049214-2.5G
101886-760EA 364.62 USD
101886-760 101886-762
1-(Tetrahydro-2H-pyran-4-ylcarbonyl)-3-piperidinecarboxylic acid
1-(Tetrahydro-2H-pyran-4-ylcarbonyl)-3-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₉NO₄
MDL Number: MFCD11170070
CAS Number: 1042799-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR