1-(Tetrahydro-2H-pyran-4-ylcarbonyl)-2-piperidinecarboxylic acid

Supplier: Matrix Scientific

049213-500MG 049213-2.5G
101886-756EA 364.62 USD
101886-756 101886-758
1-(Tetrahydro-2H-pyran-4-ylcarbonyl)-2-piperidinecarboxylic acid
1-(Tetrahydro-2H-pyran-4-ylcarbonyl)-2-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₉NO₄
MDL Number: MFCD11170085
CAS Number: 1218627-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR