1-(Tetrahydro-2H-pyran-4-ylcarbonyl)proline

Supplier: Matrix Scientific

049212-500MG 049212-2.5G
101886-752EA 364.62 USD
101886-752 101886-754
1-(Tetrahydro-2H-pyran-4-ylcarbonyl)proline
1-(Tetrahydro-2H-pyran-4-ylcarbonyl)proline

Hazard: Irritant

Formula: C₁₁H₁₇NO₄
MDL Number: MFCD11170054
CAS Number: 1308987-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR