N-(Tetrahydro-2H-pyran-4-ylcarbonyl)alanine

Supplier: Matrix Scientific

049211-500MG 049211-2.5G
101886-748EA 364.62 USD
101886-748 101886-750
N-(Tetrahydro-2H-pyran-4-ylcarbonyl)alanine
N-(Tetrahydro-2H-pyran-4-ylcarbonyl)alanine

Hazard: Irritant

Formula: C₉H₁₅NO₄
MDL Number: MFCD11170037
CAS Number: 1309036-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR