N-(4-Piperidinyl)tetrahydro-2H-pyran-4-carboxamide hydrochloride

Supplier: Matrix Scientific

049209-500MG 049209-2.5G
101886-740EA 364.62 USD
101886-740 101886-742
N-(4-Piperidinyl)tetrahydro-2H-pyran-4-carboxamide hydrochloride
N-(4-Piperidinyl)tetrahydro-2H-pyran-4-carboxamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₁ClN₂O₂
MDL Number: MFCD13559752
CAS Number: 1220031-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR