3-Pyrrolidinyl tetrahydro-2H-pyran-4-carboxylate hydrochloride

Supplier: Matrix Scientific

049207-500MG 049207-2.5G
101886-732EA 364.62 USD
101886-732 101886-734
3-Pyrrolidinyl tetrahydro-2H-pyran-4-carboxylate hydrochloride
3-Pyrrolidinyl tetrahydro-2H-pyran-4-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₈ClNO₃
MDL Number: MFCD13559750
CAS Number: 1220031-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR