2-(2-Piperidinyl)ethyl tetrahydro-2H-pyran-4-carboxylate hydrochloride

Supplier: Matrix Scientific

049205-500MG 049205-2.5G
101886-724EA 364.62 USD
101886-724 101886-726
2-(2-Piperidinyl)ethyl tetrahydro-2H-pyran-4-carboxylate hydrochloride
2-(2-Piperidinyl)ethyl tetrahydro-2H-pyran-4-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₄ClNO₃
MDL Number: MFCD13559748
CAS Number: 1219948-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR