2-(3-Piperidinyl)ethyl tetrahydro-2H-pyran-4-carboxylate hydrochloride

Supplier: Matrix Scientific

049204-500MG 049204-2.5G
101886-720EA 364.62 USD
101886-720 101886-722
2-(3-Piperidinyl)ethyl tetrahydro-2H-pyran-4-carboxylate hydrochloride
2-(3-Piperidinyl)ethyl tetrahydro-2H-pyran-4-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₄ClNO₃
MW: 277.79 g/mol
MDL Number: MFCD13559747
CAS Number: 1220031-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR