3-Azetidinyl tetrahydro-2H-pyran-4-carboxylate trifluoroacetate

Supplier: Matrix Scientific

049200-500MG 049200-2.5G
101886-704EA 364.62 USD
101886-704 101886-706
3-Azetidinyl tetrahydro-2H-pyran-4-carboxylate trifluoroacetate
3-Azetidinyl tetrahydro-2H-pyran-4-carboxylate trifluoroacetate

Hazard: Irritant

Formula: C₁₁H₁₆F₃NO₅
MDL Number: MFCD29925632
CAS Number: 1220021-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR