2-[Methyl(2-phenylacetyl)amino]acetic acid

Supplier: Matrix Scientific

049196-500MG 049196-2.5G
101886-688EA 364.62 USD
101886-688 101886-690
2-[Methyl(2-phenylacetyl)amino]acetic acid
2-[Methyl(2-phenylacetyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₃
MDL Number: MFCD09804393
CAS Number: 155256-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR