2-Methyl-N-(2-phenylacetyl)alanine

Supplier: Matrix Scientific

049195-500MG 049195-2.5G
101886-684EA 364.62 USD
101886-684 101886-686
2-Methyl-N-(2-phenylacetyl)alanine
2-Methyl-N-(2-phenylacetyl)alanine

Hazard: Irritant

Formula: C₁₂H₁₅NO₃
MW: 221.26 g/mol
MDL Number: MFCD13559740
CAS Number: 34028-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR