1-(2-Phenylacetyl)-3-piperidinecarboxylic acid

Supplier: Matrix Scientific

049194-500MG 049194-2.5G
101886-680EA 166.15 USD
101886-680 101886-682
1-(2-Phenylacetyl)-3-piperidinecarboxylic acid
1-(2-Phenylacetyl)-3-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₇NO₃
MDL Number: MFCD09814410
CAS Number: 1017381-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR