1-(2-Phenylacetyl)-2-piperidinecarboxylic acid

Supplier: Matrix Scientific

049193-500MG 049193-2.5G
101886-676EA 364.62 USD
101886-676 101886-678
1-(2-Phenylacetyl)-2-piperidinecarboxylic acid
1-(2-Phenylacetyl)-2-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₇NO₃
MDL Number: MFCD09803383
CAS Number: 1101843-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR