1-(2-Phenylacetyl)proline
Supplier: Matrix Scientific
049192-500MG
049192-2.5G
101886-672EA
364.62
USD
101886-672
101886-674
1-(2-Phenylacetyl)proline
1-(2-Phenylacetyl)proline
Hazard: Irritant
Formula:
C₁₃H₁₅NO₃ |
MDL Number:
MFCD00451541 CAS Number: 2752-38-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...