N-(2-Phenylacetyl)alanine

Supplier: Matrix Scientific

049191-500MG 049191-2.5G
101886-668EA 364.62 USD
101886-668 101886-670
N-(2-Phenylacetyl)alanine
N-(2-Phenylacetyl)alanine

Hazard: Irritant

Formula: C₁₁H₁₃NO₃
MDL Number: MFCD00114554
CAS Number: 17966-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR