2-Phenyl-N-(4-piperidinyl)acetamide hydrochloride

Supplier: Matrix Scientific

049189-500MG 049189-2.5G
101886-660EA 364.62 USD
101886-660 101886-662
2-Phenyl-N-(4-piperidinyl)acetamide hydrochloride
2-Phenyl-N-(4-piperidinyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O
MDL Number: MFCD11811770
CAS Number: 1170480-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR