Methyl (2S,4S)-4-[(2-phenylacetyl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

049188-500MG 049188-2.5G
101886-656EA 364.62 USD
101886-656 101886-658
Methyl (2S,4S)-4-[(2-phenylacetyl)oxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[(2-phenylacetyl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₈ClNO₄
MDL Number: MFCD13559739
CAS Number: 1354488-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR